missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Microscope Slides with Ground Edges

Examine objects with steadiness. Perfect for sturdy examination of objects under a microscope.

Brand:  Hirschmann™ 8210104

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 11622239

  • 595.00 € / Pack of 5000
Estimated Shipment: 10-07-2024
to see stock.
Add to basket

Description

Description

  • Ground Edges 90°C ISO 8037/l
  • With LOT number on packaging
  • CE acc. to IVD 98/79 EC
Specifications

Specifications

90°, Ground
100 x 50/Pk.
26 mm
CE acc. IVD 98/79 EC
76 x 26 mm
0.95 to 1.05 mm
76 mm
Extra White
Tropical climate packaging, best before 6 months after delivery
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers