Learn More
Merck Millex™-GP Syringe Filter, PES, 0.45 μm
Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses
Brand: Merck SLHPX13NK
1960.00 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses
Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:- Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
- Available in 0.22 μm and 0.45 μm pore sizes and two diameters to suit your application needs
- Low protein binding to minimize interaction with your sample and maximize recovery
- Polypropylene housing, offering a low level of extractables and broad chemical compatibility
Applications: Buffer Filtration, Clarification of Aqueous Solutions
Specifications
Green | |
Hydrophilic PES | |
Non-sterile | |
PP | |
Male Luer slip | |
Syringe Filter | |
13 mm |
Single-use | |
0.45 μm | |
Analytical sample prep | |
male luer slip | |
1000Pack | |
<10 mL | |
13 mm |