Learn More
Lovibond™ Minikit Hardness Total AF423 Test
Brand: Lovibond™ 414230
Code : 04
Additional Details : Weight : 0.28200kg
Specifications
1 Pack | |
4, 8 or 20 CaCO3mg/L | |
Minikit Hardness Total AF423 Test |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hardness Y / N
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : lithium hydroxide