Learn More
Emma Safety Footwear Mitchel XL Safety Shoes
Ideal for working with metal with optimal comfort, ventilation and moisture removal technology. A perfectly integrated steel toe protects feet against falling pieces of metal and other exterior threats. The Gripforce™ DUO soles withstand high temperatures and are oil and petrol resistant.
Brand: Emma Safety Footwear MM13035853
Additional Details : Weight : 3.05000kg
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- Nitrile Rubber Heat Resistant Outsole
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Full grain Nappa leather / Split leather | |
53 |
Laces | |
Mitchel XL | |
Leather | |
Steel |