missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 297

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 297 is a selective Kir3.1/3.2 (GIRK1/2) channel activator (IC50 values are 160, 887 and 914 nM for GIRK1/2, GIRK1/4 and GIRK1/3 respectively). Exhibits no effect on GIRK2, GIRK2/3, Kir2.1 and Kv7.4 channels, and has minimal effect on a panel of other ion channels, receptors and transporters. Prevents PTZ -induced epileptic seizures in mice. Brain penetrant.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | ML 297 |
| Purity | >99% |
| Molecular Weight (g/mol) | 328.32 |
| Inhibitors | ML 297 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction