missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ D-Tipper™ Pipetters

Made of precision engineered aluminum alloy and stainless steel with limited Lifetime Warranty (covering parts). Color coded body for easy volume identification and individually serialized with Calibration Certificate included.

Brand:  VistaLab™ Technologies 1157C

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11975882

  • 200.00 € / Each


Estimated Shipment
16-06-2021
Add to basket

Description

Description

  • Single-stroke “to deliver” operation
  • Built-in tip-ejector and glove guard
  • Factory calibrated to deliver specific quantities; can be easily recalibrated in the lab
  • Available in 30 fixed volumes ranging from 5-1000 μL
  • Also available as a Selectable model: 50/100/200 μL
Specifications

Specifications

±1%
Micro Pipetting/Liquid Transfer
0.80%
Axial
Silver
D-Tipper
700 µL
Not Autoclavable
Nozzle Insert, Calibration Key
Non-sterile
Limited Lifetime
Aluminum and Stainless Steel
1
Videos
SDS
Documents