missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lab Vision™ MMP-9 (92kDa Collagenase IV) Ab-5 Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-5, Mouse Monoclonal Antibody.

Brand:  Thermo Scientific™ MS817PABX

Code : A0

Additional Details : Weight : 0.80000kg

Product Code. 11818741

  • 648.00 € / 200µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Deliver accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-5, Mouse Monoclonal Antibody.

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Mouse

Clone: IIA5

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Human native 92kDa MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells or CCL4 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not optimum)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MMP-9 (92kDa Collagenase IV) Ab-5
Monoclonal
1mg/mL
Mouse
200 μL
Primary
Concentrated
Immunoprecipitation, Western Blot
IIA5
Unconjugated
Human native 92kDa MMP-9
RUO
Human, Rat
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers