Learn More
MMP Inhibitor II, Thermo Scientific™
A potent, reversible, broad range inhibitor of matrix metalloproteinases
Brand: Thermo Scientific Alfa Aesar J65638.#0
Code : NEW
Additional Details : CAS Number : 203915-59-7 Weight : 0.00005kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5 mg | |
-30°C to -10°C | |
203915-59-7 | |
513.58g/mol | |
Soluble in DMSO | |
Solid |
Off-white | |
A potent, reversible, broad range inhibitor of matrix metalloproteinases | |
C21H27N3O8S2 | |
N-Hydroxy-1,3-bis(4-methoxyphenylsulfonyl)-5,5-dimethylhexahydropyrimidine-2-carboxamide | |
Signal Transduction Reagent |