Learn More
Macherey-Nagel™ MN 652 Grade Filter Papers, Round
Wet strengthened for use in industrial applications. Macherey-Nagel™ MN 652 Grade Filter Papers, Round are creped technical filter papers with fast filtration speed for use with essential oils, gelatine, resin solutions, sugared spirits, syrups, and transformer/turbine oil regeneration.
Brand:  Macherey-Nagel™ 446018
Product Code. 10002691
  • 34.65 € / Pack of 100
Only null left
Estimated Shipment: 10-07-2025
Add to basket
Description
Description
High quality MN filter papers use cotton linters and refined pulp with a high level of α-cellulose. Cotton linters are short-fiber seed hairs from cotton seeds, which cannot be used for textile purposes, but which are highly suitable for the manufacture of soft and absorbent filter papers. In addition to cotton linters, Macherey-Nagel uses mainly pulp, which is obtained by chemical treatment of plant materials, e.g. coniferous or deciduous wood.
Specifications
Specifications
140 g/m2
Round
15sec.
MN 652
0.45 mm
185 mm
Creped, Wet-strengthened
Cellulose
Technical, industrial applications (e.g. essential oils, gelatine, resin solutions, spirits, syrups, and transformer/turbine oil regeneration)
100 Pack
Filter Paper