missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ MN Science Silicon Septa

Premium material giving up to 400 injections per septa.

Brand:  Trajan Scientific and Medical™ 0418505

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15278613

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • High mechanical strength septa - up to 400 injections per septa
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications

Specifications

MN
17 mm
48/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers