missing translation for 'onlineSavingsMsg'
Learn More

Manutan Mobile pedestal, beech two drawer, 600mm x 410mm x 600mm

Brand:  Manutan 111D014

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12837428

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Scratch-resistant melamine faced chipboard. Accepts A4 or foolscap, foolscap files are stored lengthways and A4 files are stored width ways. Easy self-assembly.
  • Beech finish
  • Two drawer
  • Dimensions (H x W x D) : 600mm x 410mm x 600mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers