Learn More
OHAUS™ Modular Blocks and Accessories
Constructed from solid anodized aluminum block.
134.00 € - 251.00 €
Specifications
For Use With (Equipment) | Dry block heater |
---|
Alert:
To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.
Product Code | Brand | Depth (Metric) Well | Diameter (Metric) Well | Type | Sample Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (Metric) Well | Diameter (Metric) Well | Type | Sample Type | Price | Quantity & Availability | ||
15712579
|
OHAUS™
30400159 |
3.91 cm | 1.1 cm | Block | 1.5mL Tube |
134.00 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||
15742589
|
OHAUS™
30400172 |
4.45 cm | 1.71 cm | Block | 15mL Tube |
134.00 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||
15702589
|
OHAUS™
30400168 |
4.76 cm | 2.9 cm | Block | 50mL Tube |
134.00 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||
15752569
|
OHAUS™
30400153 |
4.8 cm | 1.75 cm | Block | 15/16mm Tube |
134.00 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||
15732569
|
OHAUS™
30400151 |
4.8 cm | 1.07 cm | Block | 10mm Tube |
134.00 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||
15782659
|
OHAUS™
30400246 |
— | — | Temperature Probe Kit | — |
251.00 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||
Description
Modular blocks are sold separately from heaters.
- Close contact of tubes-to-block-walls allow for maximum heat retention
- Each block has a thermometer well for measuring block temperature
Specifications
Dry block heater |