Learn More
Invitrogen™ OxyBURST™ Green H2HFF BSA - Special Packaging
Sensitive fluorogenic reagent for detecting extracellular release of oxidative products in a spectrofluorometer/fluorescence microscope
Brand: Invitrogen™ O13291
Additional Details : Weight : 0.00525kg
Description
Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Supplement | |
Free Radical Sensor | |
OXYBURST™ | |
Room Temperature |
Fluorescence | |
Solid | |
Fluorescent Dye-Based | |
Fluorogenic |
For Research Use Only. Not for use in diagnostic procedures.