missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular sieves 3A, deuterated, Thermo Scientific™
73.30 € - 178.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11946971
|
Thermo Scientific Acros
432150050 |
5 g |
73.30 €
5g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11936971
|
Thermo Scientific Acros
432150250 |
25 g |
178.00 €
25g |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Drying agentRUO – Research Use Only