missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 5A, 1-2mm (0.04-0.08in) dia. pellets, Thermo Scientific Chemicals

Catalog Number p-7080392
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
69912-79-4
AlNaO6Si2
202.14
MFCD00147627
URGAHOPLAPQHLN-UHFFFAOYSA-N
19758701
aluminum;sodium;dioxido(oxo)silane
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
This item is not returnable. View return policy
69912-79-4
AlNaO6Si2
202.14
MFCD00147627
URGAHOPLAPQHLN-UHFFFAOYSA-N
19758701
aluminum;sodium;dioxido(oxo)silane
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Molecular sieves, 5A, 1-2mm (0.04-0.08in) dia. Pellets are used in desiccation and purification of air. It finds application in natural gas and liquid petroleum gas especially for dehydration and desulphurization. In chemistry laboratory, it is used for separating compounds and drying reaction starting materials. In addition, it is used as commercial adsorbents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Sensitivity Hygroscopic
Quantity 250 g
Physical Form Pellet
Solubility Information Insoluble in water and most organic solvents.
Chemical Name or Material Molecular sieves 5A, 1 to 2mm (0.04 to 0.08 in.) dia. pellets

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'