missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(IV) sulfide, 98.5%, ACROS Organics™

Brand:  Acros Organics 215780025

Code : 29

Additional Details : CAS Number : 1317-33-5 Weight : 2.55000kg


Packaging Plastic drum
Quantity 2.5kg

 View more versions of this product

Product Code. 11954221

  • 869.00 € / 2.50kg

Estimated Shipment
27-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1317-33-5
160.07
molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
CHEBI:30704
S=[Mo]=S
MoS2
CWQXQMHSOZUFJS-UHFFFAOYSA-N
14823
bis(sulfanylidene)molybdenum

Specifications

Molybdenum(IV) sulfide
98% min. (MoS2)
MoS2
molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
CWQXQMHSOZUFJS-UHFFFAOYSA-N
bis(sulfanylidene)molybdenum
14823
160.07
98.5%
Solubility in water: insoluble. Other solubilities: soluble in aqua regia and sulfuric acid
2375.0°C
1317-33-5
MoS2
15,6322
Solubility in water: insoluble. Other solubilities: soluble in aqua regia and sulfuric acid
S=[Mo]=S
160.07
CHEBI:30704
Powder
Plastic drum
Black or Gray
2.5kg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Molybdenum(IV) sulfide

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H332-Harmful if inhaled.

Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents