missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum wire, 1.58mm (0.0625in) dia, annealed, 99.9+% (metals basis), Thermo Scientific Chemicals

Catalog Number 15406705
Click to view available options
Quantity:
10 m
5 m
This item is not returnable. View return policy
7439-98-7
Mo
95.95
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
23932
CHEBI:28685
molybdenum
[Mo]
This item is not returnable. View return policy
7439-98-7
Mo
95.95
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
23932
CHEBI:28685
molybdenum
[Mo]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 m
Percent Purity ≥99.9%
Weight ≈20.2g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Molybdenum wire, 1.58mm (0.0625 in.) dia., Annealed

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'