missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MOPS, 0.5M buffer soln., pH 7.5

Catalog Number p-7042532
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
CHEBI:44115
3-morpholin-4-ylpropane-1-sulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
This item is not returnable. View return policy
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
CHEBI:44115
3-morpholin-4-ylpropane-1-sulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1

MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

Solubility
Soluble in water.

Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
TRUSTED_SUSTAINABILITY
Concentration 0.5 M
pH 7.5
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Solubility Information Soluble in water.
Chemical Name or Material MOPS buffer soln.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'