missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H2-Ab1, Mouse, Biotin, Clone: 25-9-3, Abnova™
Mouse monoclonal antibody raised against H2-Ab1.
Brand: Abnova MAB5799.500ug
Additional Details : Weight : 0.00010kg
Specifications
H2-Ab1 | |
Monoclonal | |
Biotin | |
Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
H2-Ab1 | |
H2-Ab1 | |
C3H.SW splenocytes. | |
RUO | |
14961 | |
Mouse | |
Liquid |
Flow Cytometry | |
25-9-3 | |
Mouse monoclonal antibody raised against H2-Ab1. | |
In PBS (0.09% sodium azide) | |
AI845868/Abeta/H-2Ab/H2-Ab/I-Ab/I-Abeta/IAb/Ia-2/Ia2/Rmcs1 | |
Mouse | |
500 μg | |
Primary | |
The monoclonal antibody 25-9-3 reacts specifically with the I-Ab haplotype of MHC Class II molecules. Class II antigens are predominantly expressed on antigen-presenting cells including B lymphocytes, macrophages, dendritic cells and certain epithelial cells. | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgM κ |