missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klrb1c, Mouse, APC, Clone: PK136, Abnova™
Mouse monoclonal antibody raised against native Klrb1c.
Brand: Abnova MAB5778.100ug
Additional Details : Weight : 0.00010kg
Specifications
Klrb1c | |
Monoclonal | |
APC | |
Flow Cytometry (0.5 ug/106 cells) The optimal working dilution should be determined by the end user. | |
Klrb1c | |
Klrb1c | |
Native purified Klrb1c from CE mouse spleen and bone marrow cells enriched for NK-1+ cells. | |
RUO | |
17059 | |
Mouse | |
Liquid |
Flow Cytometry, Immunoprecipitation | |
PK136 | |
Mouse monoclonal antibody raised against native Klrb1c. | |
In PBS (0.09% sodium azide) | |
AI462337/CD161/Ly-59/Ly55c/Ly59/NK-RP1/NK1.1/NKRP1/NKRP140/Nk-1/Nk-1.2/Nk1/Nk1.2/Nkrp1c | |
Mouse | |
100 μg | |
Primary | |
Mouse NK1.1 alloantigen, Mr 80-85 KDa (39 KDa under reducing conditions). | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |