missing translation for 'onlineSavingsMsg'
Learn More

RICTOR, Mouse, Clone: 1F3, Abnova™

Mouse monoclonal antibody raised against a partial recombinant RICTOR.

Brand:  Abnova H00253260-M01A.200uL

Additional Details : Weight : 0.00010kg

Product Code. 16162567

  • 326.00 € / 200µL
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RICTOR and MTOR (FRAP1; MIM 601231) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth (Sarbassov et al., 2004 [PubMed 15268862]).[supplied by OMIM

Sequence: MAAIGRGRSLKNLRVRGRNDSGEENVPLDLTREPSDNLREILQNVARLQGVSNMRKLGHLNNFTKLLCDIGHSEEKLGFHYEDIIICLRLALLNEAKE
Specifications

Specifications

RICTOR
Monoclonal
Unconjugated
ascites with no preservative
NM_152756
RICTOR
RICTOR (NP_689969,1 a.a. ∽ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
253260
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Ascites
ELISA, Western Blot
1F3
Mouse monoclonal antibody raised against a partial recombinant RICTOR.
RICTOR
DKFZp686B11164/KIAA1999/MGC39830/mAVO3
Mouse
200 μL
Primary
Human
Antibody
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers