missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG3 Isotype Control (PPV-07), Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MA110433
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody will not cross-react with human, porcine, mouse or rat.
Mouse IgG3 Control antibody specifically detects Mouse IgG3 in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation.Specifications
Mouse IgG3 | |
PPV-07 | |
Unconjugated | |
IgG3; Ighg3; Immunoglobulin G; Immunoglobulin G3; Immunoglobulin heavy constant gamma 3; ImmunoglobulinG; ImmunoglobulinG3 | |
Plum pox virus | |
100 μg | |
Isotype Control (Primary) | |
4° C, do not freeze | |
IgG3 |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
1 mg/mL | |
HEPES buffered saline with 15mM sodium azide; pH 7.0 | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |