missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ROBO4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB50041-SP
This item is not returnable.
View return policy
Description
ROBO4 Monoclonal specifically detects ROBO4 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| ROBO4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ECSM4, FLJ20798, Magic roundabout, MGC133352, MGC133353, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout (Drosophila) | |
| Y3 rat myeloid cell line transfected with mouse ROBO4 Leu28-Ser1012 Accession # Q8C310 | |
| 25 μg | |
| Primary | |
| Detects mouse ROBO4 in direct ELISAs. | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 274914 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Q8C310 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54538 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction