Learn More
Invitrogen™ Mouse VEGFR3 ELISA Kit
Mouse VEGFR3 quantitates mouse VEGFR3 in serum, plasma, supernatant.
Brand: Invitrogen™ EM73RB
Additional Details : Weight : 0.33820kg
Description
Mouse VEGFR3 quantitates mouse VEGFR3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse VEGFR3.
Vascular endothelial growth factor is a key regulator of blood vessel development in embryos and angiogenesis in adult tissues. Unlike VEGF, the related VEGFC stimulates the growth of lymphatic vessels through its specificl ymphatic endothelial receptor VEGFR-3. In 1998, Dumont showed that targeted inactivation of the VEGFR-3 gene in mice resulted in defective blood vessel development in early embryos. Vasculogenesis and angiogenesis occurred, but large vessels became abnormally organized with defective lumens, leading to fluid accumulation in the pericardial cavity and cardiovascular failure at embryonic day 9.5. Thus, VEGFR3 has an essential role in the development of theembryonic cardiovascular system before the emergence of the lymphatic vessels.Specifications
AI323512; Chy; chylous ascites; FLT4; FLT-4; FLT41; fms related tyrosine kinase 4; FMS-like tyrosine kinase 4; fms-related tyrosine kinase 4; LMPH1A; PCL; receptor protein tyrosine kinase; soluble VEGFR 3; sVEGFR 3; tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; vascular endothelial growth factor receptor-3; VEGF R3; Vegfr3; VEGFR-3 | |
0.2 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
FLT4 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.2 to 50 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P35917 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Plasma, 50 μL; Supernatant, 50 μL; Serum, 50 μL | |
VEGF Receptor 3 | |
4 hr 45 min |