Learn More
MP Biomedicals™ 1X Phosphate Buffered Saline, Dulbecco's, without Magnesium and Calcium
Clear, colorless saline solution without magnesium and calcium
Brand: MP Biomedicals™ 091860454X6
168.34 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : CAS Number : 7778-77-0 Weight : 3.00000kg
Description
- MDL Number: MFCD00131855
- Format: Solution
- pH: 7 to 7.6
- Osmolality: 270 to 305 mM/kg
For handling and culturing of mammalian cells
Specifications
Clear | |
1 X | |
6 x 500 mL | |
7 to 7.6 |
15° to 30°C | |
Phosphate Buffered Saline (PBS) | |
Potassium Chloride 200mg/L, Potassium Phosphate Monobasic 200mg/L, Sodium Chloride 8000mg/L, Sodium Phosphate Dibasic 1150mg/L | |
Liquid |