missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Albumin 35% Solution
Globulin free albumins are suitable for use in applications where no other proteins should be present
336.02 € - 3721.66 €
Specifications
Concentration | 34.4g/dL |
---|---|
For Use With (Application) | Immunoassay, Mammalian Cell Culture |
Molecular Weight (g/mol) | ∽66 |
Name | Bovine Albumin 35% Solution |
pH Range | 7.2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404216
|
MP Biomedicals™
08810061 |
50mL |
336.02 €
50mL |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11414216
|
MP Biomedicals™
08810063 |
1L |
3721.66 €
1000mL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Albumins are a group of acidic proteins which occur plentifully in the body fluids and tissues of mammals and in some plant seeds.
- CAS Number: 9048-46-8
- MDL Number: MFCD00130384
- Format: Solution
- Sterile albumins
- Concentration: About 35g/dl
For immunoassays and mammalian cell culture. Used to solubilize lipids and is also used as a blocking agent in Western blots or ELISA applications.
Specifications
34.4g/dL | |
∽66 | |
7.2 | |
Solution |
Immunoassay, Mammalian Cell Culture | |
Bovine Albumin 35% Solution | |
0° to 5°C |