Learn More
MP Biomedicals™ Ampicillin, Trihydrate
Serves a variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Brand: MP Biomedicals™ 0219014825
133.36 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 69-52-3 Weight : 0.02500kg
Description
Alll antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Applications:
Selection antibiotic effective against most gram negative and gram positive bacteria.
Specifications
4°C | |
Antibiotic | |
25 g |
Powder | |
8 to 10 |
Safety and Handling
- Ampicillin Sodium Salt
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.