Learn More
MP Biomedicals™ Casein, Hammerstein Grade
Casein is the name for a family of related phosphoproteins
Brand: MP Biomedicals™ 0210128901
Code : NEW
Additional Details : CAS Number : 9000-71-9 Weight : 2.26800kg
Description
Partial gastrointestinal digestion of casein is a rich source of bioactive peptides, such as β-casomorphin.
- CAS Number: 9000-71-9
- EC Number: 232-555-1
- MDL Number: MFCD00081481
- Molecular Weight: 75,000 to 100,000
- Format: white powder
- Soluble in alkaline aqueous solutions; very sparingly soluble in water and non-polar organic solvents
- Moisture content: 6±2%
- pH: 5.5 (2% suspension)
Used in production and characterization of phage-display recombinant antibody against carrageenans.
Specifications
75000 to 100000 | |
5.5 | |
15° to 30°C |
Casein | |
1 lb. | |
Powder |