missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Fetal Bovine Serum, CellECT™ Silver

Aseptically collected via cardiac puncture

Brand:  MP Biomedicals™ 092916254

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11441465

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Collected from a USDA regulated and monitored abbatoir by technicians who work onsite
  • Careful handling minimizes cell lysis and other detrimental factors, thereby reducing possible lysozyme and hemoglobin contamination
  • Supplied sterile, with no detectable mycoplasma or viral infection
  • Endotoxin level: slightly more than 1 ng/mL
Specifications

Specifications

1.02g/mL
Golden Brown
USDA
Yes
Used to supplement cell culture media for the optimal growth of many cell types, effective in promoting and sustaining growth of vertebrate mammalian and insect cells
280 to 325mOsm/kg
6.7 to 7.8
Heat Inactivated
USDA Inspected
Sterile
3.1 to 4.4g/dL
Slightly exceed 1ng/mL
Liquid
27mg/dL
No
-20°C
1 x 500mL
Animal Serum Culture Media
Videos
Documents