Learn More
MP Biomedicals™ Geneclean™ for Ancient DNA Kit
Purifies fragmented or damaged DNA from aged or preserved samples
Brand: MP Biomedicals™ 111002200
Code : 50
Additional Details : Weight : 0.20000kg Transport : UN number : 2922 Chem class : 8 Pack group : II
Includes:
- Dehybernation Solutions A, A2 and B
- Ancient DNA GLASSMILK
- Salton Wash £1 and £2
- Ancient Alcohol Wash
- DNA-free Elution Solution
- Spin filter with tube
- Catch tube
- Lysing Matrix K
Description
- Format: Geneclean™ slurry plus SPIN filter
- Size: 100 preps
Specifications
Geneclean Slurry Plus Spin Filter | |
Dehybernation solution A, B, A2, Ancient DNA Glassmilk, Ancient DNA Alcohol wash, DNA-free elution solution, Homogenization matrix | |
Ancient DNA Kit |
Purifies fragmented or damaged DNA from aged or preserved samples | |
100 Preps |
Safety and Handling
- geneclean for ancient DNA kit
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : guanidine thiocyanate, propietary reagent