Learn More
MP Biomedicals™ Serum, Type AB, Human (HCV, HIV, HbsAG Free)
Used as a growth supplement for cell culture media and in other research applications.
Brand: MP Biomedicals™ 092931949
Code : NEW
Additional Details : Weight : 0.14400kg
Description
- Collected from type AB human blood
- Used as a growth supplement for cell culture media and in other research applications
- Proven to grow many human cell lines at a faster rate than mixed blood group serum
Specifications
U.S. regulations | |
-20°C | |
Liquid | |
100 mL | |
0.1 μm filtration | |
≤26 mg/dL | |
263 to 323 mOsm/kg | |
5.5 to 7.5 g/dL |
Golden Brown | |
Human | |
Human Serum | |
Sterile | |
Grow many Human cell lines at a faster rate | |
No | |
7 to 8 |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.