Learn More
MP Biomedicals™ Serum, Rabbit
Supports cultures of Borrelia burgdorferi, the causative agent of Lyme disease.
Brand: MP Biomedicals™ 092941149
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This serum is derived from the blood of healthy adult animals. Rabbit serum is used to support cultures of Borrelia burgdorferi, the causative agent of Lyme disease.Specifications
U.S. regulations | |
-20°C | |
Frozen | |
100 mL | |
Sterile | |
Used to support cultures of Borrelia burgdorferi, the causative agent of Lyme disease | |
No | |
7.4 to 7.9 |
Dark Brown | |
Rabbit | |
Rabbit Serum | |
4 to 6.6 g/dL | |
Filtration | |
≤33 mg/dL | |
268 to 326 mOsm/kg |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.