missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ 0.05% Trypsin-EDTA (1X)
0.05% Trypsin and 0.5mM EDTA in modified HBSS w/o Ca++ and Mg++, 100 mL
Brand: MP Biomedicals™ 091689149
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Concentration: 0.05% (w/v) Trypsin (1:250) and 0.02% (w/v) EDTA (Versene)Specifications
Cell dissociation, routine cell culture passaging and primary tissue dissociation due to its digestive strength | |
Porcine | |
Solution | |
Trypsin-EDTA |
7 to 7.4 | |
0°C to 20°C | |
100 mL |