missing translation for 'onlineSavingsMsg'
Learn More

Welch™ MPC 601 Tp ATEX Cat.2 Diaphragm Pump

Brand:  Welch™ 4000511-04

Code : 04

Additional Details : Weight : 30.00000kg

Product Code. 12955134

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

75 L/min.
425 mm
< 44 dB
60°C
50 Hz
240 mm
1.5 bar
400 V 50 Hz
MPC 601 Tp ATEX Cat.2
29.7 kg
10°C
272 mm
Diaphragm Pump
400 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers