missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent

Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.

Brand:  Thermo Scientific™ TS-48914

Code : 16

Additional Details : CAS Number : 24589-78-4 Weight : 0.10000kg Transport : UN number : 1993 Chem class : 3 Pack group : III


 View more versions of this product

Product Code. 11507680

  • 692.39 € / Each
Estimated Shipment: 16-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Use Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent for GC analyses of early eluting compounds that would otherwise be obscured in the chromatogram. MSTFA, the most volatile TMS-amide available, and its byproduct N-methyltrifluoroacetamide typically elute with the solvent front, allowing TMS derivatives of small molecules to be analyzed. Addition of Thermo Scientific™ TMCS aids derivatization of amides, secondary amines, and hindered hydroxyls not derivatized by MSTFA alone.

  • Trimethylsilyl donor strength comparable to BSA and BSTFA
  • Reacts to replace labile hydrogens on a wide range of polar compounds with a Si(CH3)3 group
  • Used to prepare volatile and thermally stable derivatives for GC and MS
  • Volatile byproduct, N-methyltrifluoroacetamide, has an even lower retention time than MSTFA
  • Often TMS derivatives of small molecules can be analyzed when derivatized with MSTFA because the byproducts and the reagent itself usually elute with the solvent front
  • Addition of Thermo Scientfic TMCS aids derivatization of amides, secondary amines and hindered hydroxyls not derivatized by MSTFA alone
Specifications

Specifications

GC/MS
Flammable, Irritant
100 mL
98 to 100%
2°C to 8°C
MSTFA (N-Methyl-N-trimethylsilyltrifluoroacetamide)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MSTFA, 100 mL. N-Methyl-N-TMS-Trifluoro

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P370+P378-In case of fire: Use to extinguish.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.