missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HUBER Metal Hose MT-400-1-M24x1,5
Brand: HUBER 6784
Code : NEW
Additional Details : Weight : 0.80000kg
Description
Special insulation with additional smooth silicon hose, ends covered with silicon capsSpecifications
M24 x 1.5 Thread | |
1000 mm | |
0.8 kg |
Metal | |
1 Ea. | |
Hose |