missing translation for 'onlineSavingsMsg'
Learn More

Mucic Acid 98%, Acros Organics

Brand:  Acros Organics 128041000

Code : 29

Additional Details : CAS Number : 526-99-8 Weight : 0.15000kg

Packaging Plastic bottle
Quantity 100g

 View more versions of this product

Product Code. 10428480

  • 31.50 € / 100g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

526-99-8
210.138
DSLZVSRJTYRBFB-DUHBMQHGSA-N
3037582
C6H10O8
MFCD00004239
Galactaric acid
CHEBI:30852

Specifications

Mucic acid
526-99-8
100.0
≥97.5%
C6H10O8
MFCD00004239
15, 4361
Galactaric acid
C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O
210.138
CHEBI:30852
Fine Crystalline Powder
White to Beige
98%
97.5
Authentic
98%
0.5% max.
03, IV, 1292
Plastic bottle
DSLZVSRJTYRBFB-DUHBMQHGSA-N
(2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid
3037582
210.14
0.2% max.
100g
Videos
SDS