Learn More
Agilent BioTek MultiFlo FX Wash Module
Wash module for 6- to 384-well plates combines dispensing and washing automated on one platform
Brand: Agilent BioTek 1260023
Additional Details : Weight : 12.00000kg
Specifications
96-/384-well plates, 300 μL/well: <3% CV,6-well plates, 5560 μL/well: <5% CV | |
Primary/Secondary Screening Assays,Compound Storage,Genomics and Proteomics Research,ELISA,Cell Based Washing, Fixing, Staining, Volume or Concentration Normalization | |
Wash module, pump, bottles, tubing, 96/384 wash manifold |
External wash module | |
CE and TUV marked. RoHS compliant. | |
MFXP2, MFXP2R |