Learn More
Thermo Scientific™ Microplate Reader Verification Plates
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Brand: Thermo Scientific™ 24072800
Code : A0
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Features:- Wide wavelength range of 340 to 850nm
- Fast reading of both 96- and 384-well plates
- Shaking and incubation up to 50°C for temperature critical assays
- Ease of use through the large color screen and a variety of language versions
- Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
- USB port for easy data transfer
Recommended for:
Immunoassays (ELISA), protein assays, endotoxins, cytotoxicity and proliferation assays, enzyme assays, growth curves
Specifications
Multiskan Verification Plate, 8 wavelengths | |
Verification Plate | |
NPL (National Physical Laboratory) traceable Multiskan™ Verification Plate Certificate of Calibration USB memory device including SkanIt™ software sessions and reference files Carrying case for Multiskan™ Verification Plate |
Multiskan FC | |
Multiskan™ |
For Research Use Only. Not for use in diagnostic procedures.