missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,3-Dichlorophenyl)-2-fluorobenzamide, 97%

Brand:  Alfa Aesar™ H59608.03

Code : NEW

Additional Details : CAS Number : 200710-81-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415988

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,3-Dichlorophenyl)-2-fluorobenzamide
200710-81-2
284.12
MFCD00028249

Specifications

N-(2,3-Dichlorophenyl)-2-fluorobenzamide
Solid
284.12
1g
200710-81-2
97%
MFCD00028249
Videos
SDS