missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Dibromophenyl)-2-chlorobenzamide, 97%

Brand:  Alfa Aesar™ H59144.03

Code : NEW

Additional Details : CAS Number : 339252-38-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415368

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 20-11-2018
Add to basket

Specifications

Chemical Identifiers

339252-38-9
389.471
MMZBFOGTVYPDDW-UHFFFAOYSA-N
1513583
C1=CC=C(C(=C1)C(=O)NC2=C(C=C(C=C2)Br)Br)Cl
C13H8Br2ClNO
MFCD00820270
2-chloro-n-2,4-dibromophenyl benzamide, n-2,4-dibromophenyl-2-chlorobenzamide
2-chloro-N-(2,4-dibromophenyl)benzamide

Specifications

N-(2,4-Dibromophenyl)-2-chlorobenzamide
339252-38-9
MFCD00820270
MMZBFOGTVYPDDW-UHFFFAOYSA-N
2-chloro-N-(2,4-dibromophenyl)benzamide
1513583
Solid
1g
C13H8Br2ClNO
2-chloro-n-2,4-dibromophenyl benzamide, n-2,4-dibromophenyl-2-chlorobenzamide
C1=CC=C(C(=C1)C(=O)NC2=C(C=C(C=C2)Br)Br)Cl
389.471
389.47
97%
Videos
SDS