missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,5-Difluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60428.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416898

  • 646.00 € / 1g


Estimated Shipment date
from Supplier 10-12-2018
Add to basket

Specifications

Chemical Identifiers

109023-72-5
269.27
QKPONFDTNDFKFQ-UHFFFAOYSA-N
8500589
C12H9F2NO2S
MFCD05136551
n-2,5-difluorophenyl benzenesulfonamide, qkponfdtndfkfq-uhfffaoysa-n, 2,5-difluoro-n-benzenesulphonylaniline, 2,5-difluorophenyl phenylsulfonyl amine

Specifications

N-(2,5-Difluorophenyl)benzenesulfonamide
1g
C12H9F2NO2S
15129639
QKPONFDTNDFKFQ-UHFFFAOYSA-N
8500589
Solid
Cream to White
109023-72-5
MFCD05136551
n-2,5-difluorophenyl benzenesulfonamide, qkponfdtndfkfq-uhfffaoysa-n, 2,5-difluoro-n-benzenesulphonylaniline, 2,5-difluorophenyl phenylsulfonyl amine
269.27
269.27
97%
Videos
SDS