missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[2-Chloro-5-(trifluoromethyl)phenyl]-3,4-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61441.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416516

  • 119.52 € / 1g


Estimated Shipment date
from Supplier 20-11-2018
Add to basket

Specifications

Chemical Identifiers

871923-03-4
371.71
UNYKFOQQQVERSO-UHFFFAOYSA-N
5016140
C13H7ClF5NO2S
MFCD05116126
n-2-chloro-5-trifluoromethyl phenyl-3,4-difluorobenzene-1-sulfonamide, n-2-chloro-5-trifluoromethyl phenyl-3,4-difluorobenzenesulfonamide

Specifications

N-[2-Chloro-5-(trifluoromethyl)phenyl]-3,4-difluorobenzenesulfonamide
871923-03-4
MFCD05116126
UNYKFOQQQVERSO-UHFFFAOYSA-N
5016140
Solid
1g
C13H7ClF5NO2S
n-2-chloro-5-trifluoromethyl phenyl-3,4-difluorobenzene-1-sulfonamide, n-2-chloro-5-trifluoromethyl phenyl-3,4-difluorobenzenesulfonamide
371.71
371.71
97%
Videos
SDS