missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3,5-Dichlorophenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59279.03

Code : NEW

Additional Details : CAS Number : 316132-04-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415548

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(3,5-Dichlorophenyl)-3-methylbenzamide
316132-04-4
280.15
MFCD01047342

Specifications

N-(3,5-Dichlorophenyl)-3-methylbenzamide
Solid
280.15
1g
316132-04-4
97%
MFCD01047342
Videos
SDS