missing translation for 'onlineSavingsMsg'
Learn More

N-(3'-Aminopropyl)-2-pyrrolidinone, 95%, ACROS Organics™

Brand:  Acros Organics 159600050

Code : 29

Additional Details : CAS Number : 7663-77-6 Weight : 0.05500kg Transport : UN number : 3267 Chem class : 8 Pack group : II


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10426820

  • 28.00 € / 5g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

7663-77-6
142.202
HJORCZCMNWLHMB-UHFFFAOYSA-N
82111
C1CC(=O)N(C1)CCCN
C7H14N2O
MFCD00003201
1-3-aminopropyl pyrrolidin-2-one, n-3'-aminopropyl-2-pyrrolidinone, 2-pyrrolidinone, 1-3-aminopropyl, 1-3-aminopropyl-2-pyrrolidone, n-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, 1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, zlchem 943
1-(3-aminopropyl)pyrrolidin-2-one

Specifications

N-(3'-Aminopropyl)-2-pyrrolidinone
>112°C
Glass bottle
120.0°C to 123.0°C (1.0 mmHg)
5g
94.0
95%
MFCD00003201
HJORCZCMNWLHMB-UHFFFAOYSA-N
1-(3-aminopropyl)pyrrolidin-2-one
82111
Liquid
1.0100g/mL
Authentic
1.4970 to 1.4990
Colorless to Brown
7663-77-6
100.0
C7H14N2O
1-3-aminopropyl pyrrolidin-2-one, n-3'-aminopropyl-2-pyrrolidinone, 2-pyrrolidinone, 1-3-aminopropyl, 1-3-aminopropyl-2-pyrrolidone, n-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, 1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, zlchem 943
C1CC(=O)N(C1)CCCN
142.202
142.2
95%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-(3'-Aminopropyl)-2-pyrrolidinone

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS