missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Fluoro-4-bromophenyl)-2-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59331.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415608

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Fluoro-4-bromophenyl)-2-methoxybenzamide
MFCD20807199

Specifications

N-(3-Fluoro-4-bromophenyl)-2-methoxybenzamide
Solid
MFCD20807199
324.15
97%
1g
Videos
SDS