missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Fluorophenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59862.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416298

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 03-09-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Fluorophenyl)-3,5-dinitrobenzamide
MFCD00407313

Specifications

N-(3-Fluorophenyl)-3,5-dinitrobenzamide
MFCD00407313
305.22
97%
1g
Solid
Videos
SDS