Learn More
N-(3-Formyl-4-pyridinyl)-2,2-dimethylpropanamide, 97%, Maybridge™
Brand: Maybridge MO00095DA
Code : 31
Additional Details : CAS Number : 86847-71-4 Weight : 0.05100kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
86847-71-4 | |
206.245 | |
ICMXCEJBHWHTBH-UHFFFAOYSA-N | |
2779664 | |
CC(C)(C)C(=O)NC1=C(C=NC=C1)C=O |
C11H14N2O2 | |
MFCD03086208 | |
n-3-formylpyridin-4-yl pivalamide, n-3-formyl-4-pyridinyl-2,2-dimethylpropanamide, n-3-formylpyridin-4-yl-2,2-dimethylpropanamide, n-3-formyl-pyridin-4-yl-2,2-dimethyl-propionamide, 4-2,2-dimethylpropanoyl amino pyridine-3-carboxaldehyde, 2,2-dimethyl-n-3-formylpyridin-4-yl propanamide, 4-2,2-dimethylpropanoyl amino nicotinaldehyde, propanamide,n-3-formyl-4-pyridinyl-2,2-dimethyl, 4-2,2-dimethylpropanoyl amino-3-formylpyridine, n-3-formylpyrdn-4-yl pvalamde | |
N-(3-formylpyridin-4-yl)-2,2-dimethylpropanamide |
Specifications
N-(3-Formyl-4-pyridinyl)-2, 2-dimethylpropanamide | |
1g | |
97.0 | |
C11H14N2O2 | |
n-3-formylpyridin-4-yl pivalamide, n-3-formyl-4-pyridinyl-2,2-dimethylpropanamide, n-3-formylpyridin-4-yl-2,2-dimethylpropanamide, n-3-formyl-pyridin-4-yl-2,2-dimethyl-propionamide, 4-2,2-dimethylpropanoyl amino pyridine-3-carboxaldehyde, 2,2-dimethyl-n-3-formylpyridin-4-yl propanamide, 4-2,2-dimethylpropanoyl amino nicotinaldehyde, propanamide,n-3-formyl-4-pyridinyl-2,2-dimethyl, 4-2,2-dimethylpropanoyl amino-3-formylpyridine, n-3-formylpyrdn-4-yl pvalamde | |
CC(C)(C)C(=O)NC1=C(C=NC=C1)C=O | |
206.245 | |
206.24 |
Amber Glass Bottle | |
86847-71-4 | |
97% | |
MFCD03086208 | |
ICMXCEJBHWHTBH-UHFFFAOYSA-N | |
N-(3-formylpyridin-4-yl)-2,2-dimethylpropanamide | |
2779664 | |
97% |