missing translation for 'onlineSavingsMsg'
Learn More

n-(3-Formylphenyl)methanesulfonamide, ≥97%, Maybridge™

Brand:  Maybridge MO07105DA

Code : 31

Additional Details : CAS Number : 55512-05-5 Weight : 0.05100kg

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10004982

  • 129.00 € / 1g

Estimated Shipment
04-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

55512-05-5
199.224
CBDSSTWZEANOCR-UHFFFAOYSA-N
2794787
CS(=O)(=O)NC1=CC=CC(=C1)C=O
C8H9NO3S
MFCD03198179
n-3-formylphenyl methanesulfonamide, 3-methylsulfonamido benzaldehyde, methanesulfonamide, n-3-formylphenyl, 3-methylsulfonyl amino benzaldehyde, n-3-formylphenyl methanesulphonamide, m-methylsulfonylamino benzaldehyde, 3-methylsulfonylamino benzaldehyde, methanesulfonamide,n-3-formylphenyl, n-3-formyl-phenyl-methanesulfonamide
N-(3-formylphenyl)methanesulfonamide

Specifications

N-(3-formylphenyl)methanesulfonamide
1g
97.0
C8H9NO3S
n-3-formylphenyl methanesulfonamide, 3-methylsulfonamido benzaldehyde, methanesulfonamide, n-3-formylphenyl, 3-methylsulfonyl amino benzaldehyde, n-3-formylphenyl methanesulphonamide, m-methylsulfonylamino benzaldehyde, 3-methylsulfonylamino benzaldehyde, methanesulfonamide,n-3-formylphenyl, n-3-formyl-phenyl-methanesulfonamide
CS(=O)(=O)NC1=CC=CC(=C1)C=O
199.224
199.23
Amber Glass Bottle
55512-05-5
97%
MFCD03198179
CBDSSTWZEANOCR-UHFFFAOYSA-N
N-(3-formylphenyl)methanesulfonamide
2794787
97%
Videos
SDS Documents