missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromo-2-chlorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59047.03

Code : NEW

Additional Details : CAS Number : 902091-22-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415228

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromo-2-chlorophenyl)benzamide
902091-22-9
310.58
MFCD04805165

Specifications

N-(4-Bromo-2-chlorophenyl)benzamide
Solid
310.58
1g
902091-22-9
97%
MFCD04805165
Videos
SDS